4-Chloro-6,7-dimethoxyquinazoline ≥98.0% (by HPLC, total nitrogen)
Leverantör: TCI EUROPE
|
Varning
|
C2574-5G
C2574-25G
TCIAC2574-5GEA
615
SEK
TCIAC2574-5G
TCIAC2574-25G
4-Chloro-6,7-dimethoxyquinazoline ≥98.0% (by HPLC, total nitrogen)
4-Chloro-6,7-dimethoxyquinazoline
Formel:
C₁₀H₉ClN₂O₂ MW: 224,65 g/mol |
MDL Number:
MFCD01570172 CAS nummer: 13790-39-1 |
Specifikation testresultat
Appearance | Light orange to Yellow to Green powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (with Total Nitrogen) | min. 98.0 % |
Melting point | 183.0 to 190.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...