Benzyl chloromethyl ether 75%
Leverantör: Thermo Scientific
Fara
Synonyms: ((Chloromethoxy)methyl)benzene
376950100 376950500 376952500
ACRO376950100EA 575 SEK
ACRO376950100 ACRO376950500 ACRO376952500
Benzyl chloromethyl ether 75%
Benzyl chloromethyl ether
Formel: C₈H₉ClO
MW: 156,61 g/mol
Kokpunkt: 102 °C (14 mmHg)
Densitet: 1,13 g/cm³
Flampunkt: 91 °C
Storage Temperature: Kylskåp
MDL Number: MFCD00000886
CAS nummer: 3587-60-8
UN: 2810
ADR: 6.1,III
Order Now
Specifikation testresultat
Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
GC ≥70 %