3-Hydroxy-o-toluic acid ≥97%
Leverantör: Thermo Scientific
Varning
Synonyms: 3-Hydroxy-2-methylbenzoic acid
L13743.06 L13743.03
L13743.06EA 2500 SEK
L13743.06 L13743.03
3-Hydroxy-o-toluic acid ≥97%
3-Hydroxy-o-toluic acid
Purity: 97% CAS No.: 603-80-5 Molecular Formula: C8H8O3 Formula Weight: 152.15 Odour: Odourless Melting Point: 142-146° Beilstein Reference: 2613791 MDL No.: MFCD00671541
Formel: C₈H₈O₃
MW: 152,15 g/mol
Smältpunkt: 142…146 °C
Storage Temperature: Omgivande
MDL Number: MFCD00671541
CAS nummer: 603-80-5