2-Bromo-3-nitrotoluene ≥98.0%
Leverantör: TCI
Varning
B3712-1G B3712-5G
TCIAB3712-1GEA 455 SEK
TCIAB3712-1G TCIAB3712-5G
2-Bromo-3-nitrotoluene ≥98.0%
2-Bromo-3-nitrotoluene
Formel: C₇H₆BrNO₂
MW: 216.0300 g/mol
Kokpunkt: 136°C
MDL Number: MFCD00134555
CAS nummer: 41085-43-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a0e44f8e91a94bd26e1084614ad33be5f9967893&vid=a0e44f8e91a94bd26e1084614ad33be5f9967893&prid=web&clid=SAPDAM
Order Now
Specifikation testresultat
Appearance Light orange to Yellow to Green powder to crystal
Purity (GC) min. 98.0 %
Melting point 38.0 to 42.0 deg-C