2,6-Dibromo-4-nitrotoluene 95%
Leverantör: Thermo Fisher Scientific
Varning
Synonyms: 1,3-Dibromo-2-methyl-5-nitrobenzene
L13102.06 L13102.14
L13102.06EA 1325 SEK
L13102.06 L13102.14
2,6-Dibromo-4-nitrotoluene 95%
2,6-Dibromo-4-nitrotoluene
Purity: 95% CAS No.: 110127-07-6 Molecular Formula: C7H5Br2NO2 Formula Weight: 294.94 Melting Point: 54-57° Beilstein Reference: 3259701
Formel: C₇H₅Br₂NO₂
MW: 294.94 g/mol
Smältpunkt: 54…57 °C
Storage Temperature: Omgivande
MDL Number: MFCD00151807
CAS nummer: 110127-07-6