2-Methoxythioanisole ≥98.0%

Leverantör: TCI
Varning

Synonyms: 2-(Methylsulphanyl)anisole

M1851-25G M1851-5G
TCIAM1851-25GEA 1900 SEK
TCIAM1851-25G TCIAM1851-5G
2-Methoxythioanisole ≥98.0%
2-Methoxythioanisole
Formel: C₈H₁₀OS
Kokpunkt: 142°C
Smältpunkt: 19°C
MDL Number: MFCD00792424
CAS nummer: 2388-73-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=56aa7c2687651566c5bd14bb99d8e27dcdc1f6ae&vid=56aa7c2687651566c5bd14bb99d8e27dcdc1f6ae&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR