Isoamyl-4-hydroxybenzoate ≥98.0% (by GC, titration analysis)

Leverantör: TCI EUROPE
Varning

H0214-10G H0214-25G
TCIAH0214-10GEA 630 SEK
TCIAH0214-10G TCIAH0214-25G
Isoamyl-4-hydroxybenzoate ≥98.0% (by GC, titration analysis)
Isoamyl-4-hydroxybenzoate
Formel: C₁₂H₁₆O₃
Kokpunkt: 242°C
MDL Number: MFCD00017502
CAS nummer: 6521-30-8
EINECS: 229-409-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=69a95a5324c257910606b3c557ffecc61f299ff4&vid=69a95a5324c257910606b3c557ffecc61f299ff4&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %
Melting point 60.0 to 64.0 deg-C
Solubility in Methanol almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR