2-Bromo-1,3-dimethoxybenzene ≥98%
Leverantör: Thermo Scientific
Varning
B25485.14 B25485.06 B25485.03
B25485.14EA 11675 SEK
B25485.14 B25485.06 B25485.03
2-Bromo-1,3-dimethoxybenzene ≥98%
2-Bromo-1,3-dimethoxybenzene
Purity: 98% CAS No.: 16932-45-9 Molecular Formula: C8H9BrO2 Formula Weight: 217.06 Melting Point: 91-94° MDL No.: MFCD07780177
Formel: C₈H₉BrO₂
MW: 217,06 g/mol
Smältpunkt: 91…94 °C
Storage Temperature: Omgivande
MDL Number: MFCD07780177
CAS nummer: 16932-45-9