Dicyclohexylphenylphosphine ≥95.0%

Leverantör: TCI EUROPE
Varning

Synonyms: Dicyclohexyl(phenyl)phosphine

D2411-1G D2411-5G
TCIAD2411-1GEA 645 SEK
TCIAD2411-1G TCIAD2411-5G
Dicyclohexylphenylphosphine ≥95.0%
Dicyclohexylphenylphosphine
Formel: C₁₈H₂₇P
MW: 274,38 g/mol
MDL Number: MFCD00003854
CAS nummer: 6476-37-5
EINECS: 229-334-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=73529719e81e9aa56eca1d97615413cef7908220&vid=73529719e81e9aa56eca1d97615413cef7908220&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance White to Almost white powder to crystaline
Purity (GC) min. 95.0 %
Melting point 57.0 to 61.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR