3-Fluorosalicylaldehyde ≥98.0%

Leverantör: TCI EUROPE
Varning

Synonyms: 3-Fluoro-2-hydroxybenzaldehyde

F0625-1G F0625-5G
TCIAF0625-1GEA 1050 SEK
TCIAF0625-1G TCIAF0625-5G
3-Fluorosalicylaldehyde ≥98.0%
3-Fluorosalicylaldehyde
Formel: C₇H₅FO₂
MW: 140,12 g/mol
MDL Number: MFCD00003319
CAS nummer: 394-50-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1d27509177559cf965055f8b771a89319b46c071&vid=1d27509177559cf965055f8b771a89319b46c071&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance White to Light yellow to Dark green powder to crystal
Purity (GC) min. 98.0 %
Melting point 68.0 to 72.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR