Zirconium carbide ≥99.5% (max. 200 ppm Hf)
Leverantör: Thermo Fisher Scientific
Varning
35808.18 35808.30
35808.18EA 2250 SEK
35808.18 35808.30
Zirconium carbide ≥99.5% (max. 200 ppm Hf)
Zirconium carbide
Purity: 99.5% CAS No.: 12070-14-3 Molecular Formula: ZrC Formula Weight: 103.23 Physical Form: -325 Mesh Powder Density: 6.73 Appearance: Grey Odour: Odourless Melting Point: 3540° UN No.: 3178 MDL No.: MFCD00049640
Formel: CZr
MW: 103.235 g/mol
Smältpunkt: 3540 °C
Densitet: 6.73
Storage Temperature: Omgivande
MDL Number: MFCD00049640
CAS nummer: 12070-14-3
EINECS: 235-125-1
UN: 3178
ADR: 4.1 III