2-Fluoroisonicotinic acid ≥98%

Leverantör: Thermo Fisher Scientific

Synonyms: 2-Fluoropyridine-4-carboxylic acid

44755.03
44755.03EA 4225 SEK
44755.03
2-Fluoroisonicotinic acid ≥98%
2-Fluoroisonicotinic acid

Purity: 98% CAS No.: 402-65-3 Molecular Formula: C6H4FNO2 Formula Weight: 141.10 Physical Form: Powder Melting Point: 195-497° MDL No.: MFCD02181194

Formel: C₆H₄FNO₂
MW: 141.10 g/mol
Smältpunkt: 195…497 °C
Storage Temperature: Omgivande
MDL Number: MFCD02181194
CAS nummer: 402-65-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR