4-Methyl-1H-pyrazole ≥98.0%
Leverantör: TCI EUROPE
Varning
Synonyms: Fomepizole
M0774-1G M0774-5G
TCIAM0774-1GEA 215 SEK
TCIAM0774-1G TCIAM0774-5G
4-Methyl-1H-pyrazole ≥98.0%
4-Methyl-1H-pyrazole
Formel: C₄H₆N₂
MW: 82,11 g/mol
Kokpunkt: 207°C
MDL Number: MFCD00005245
CAS nummer: 7554-65-6
EINECS: 231-445-0
Merck Index: 14,04226
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d262f283ed5bd7cc8eb070dc8fab3947c5cad491&vid=d262f283ed5bd7cc8eb070dc8fab3947c5cad491&prid=web&clid=SAPDAM
Order Now
Specifikation testresultat
Appearance White to Light yellow powder to lump
Purity (GC) min. 98.0 %