cis-2,6-Dimethylpiperidine ≥99.0% (by GC, titration analysis)
Leverantör: TCI EUROPE
Fara
Synonyms: cis-2,6-Lupetidine
D1198-25ML D1198-500ML D1198-100ML
TCIAD1198-25MLEA 405 SEK
TCIAD1198-25ML TCIAD1198-500ML TCIAD1198-100ML
cis-2,6-Dimethylpiperidine ≥99.0% (by GC, titration analysis)
cis-2,6-Dimethylpiperidine
Formel: C₇H₁₅N
MW: 113,20 g/mol
Kokpunkt: 127°C
Flampunkt: 12°C
MDL Number: MFCD00066304
CAS nummer: 766-17-6
UN: 2733
ADR: 3,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f46df621dbd8b54f2a4de2afe0b4559157712a31&vid=f46df621dbd8b54f2a4de2afe0b4559157712a31&prid=web&clid=SAPDAM
Order Now
Specifikation testresultat
Appearance Colorless to Light yellow to Light orange clear liquid
Purity (GC) min. 99.0 %
Purity (Nonaqueous Titration) min. 99.0 %