3,3-Dimethylpiperidine 95%
Leverantör: Thermo Scientific
Fara
H25753.03 H25753.MD H25753.06
H25753.03EA 7675 SEK
H25753.03 H25753.MD H25753.06
3,3-Dimethylpiperidine 95%
3,3-Dimethylpiperidine
Purity: 95% CAS No.: 1193-12-0 Molecular Formula: C7H15N Formula Weight: 113.20 Flash Point: 26°(79°F) Refractive Index: 1.4475 Odour: Amine-like Boiling Point: 134-136° UN No.: 2733 MDL No.: MFCD00005995
Formel: C₇H₁₅N
MW: 113,20 g/mol
Kokpunkt: 134…136 °C
Densitet: 0,81
Flampunkt: 26 °C (79 °F)
Storage Temperature: Omgivande
MDL Number: MFCD00005995
CAS nummer: 1193-12-0
EINECS: 214-767-6
ADR: 3,III