1,2-Pentadiene ≥95.0%

Leverantör: TCI
Fara

P0805-1G P0805-5G
TCIAP0805-1GEA 3800 SEK
TCIAP0805-1G TCIAP0805-5G
1,2-Pentadiene ≥95.0%
1,2-Pentadiene
Formel: C₅H₈
Kokpunkt: 45°C
Storage Temperature: Kylskåp
MDL Number: MFCD00039943
CAS nummer: 591-95-7
EINECS: 209-737-4
UN: 3295
ADR: 3,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=25d50b58a1cdb93c3a45c2b0a288f0f0a1bae5be&vid=25d50b58a1cdb93c3a45c2b0a288f0f0a1bae5be&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 95.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR