2-Methyl-1,4-pentadiene ≥99.0%

Leverantör: TCI EUROPE
Fara

M0616-1ML M0616-5ML
TCIAM0616-1MLEA 1200 SEK
TCIAM0616-1ML TCIAM0616-5ML
2-Methyl-1,4-pentadiene ≥99.0%
2-Methyl-1,4-pentadiene
Formel: C₆H₁₀
Kokpunkt: 57°C
Flampunkt: -3°C
MDL Number: MFCD00008601
CAS nummer: 763-30-4
EINECS: 212-109-2
UN: 2461
ADR: 3,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5b566036df87bda7bbe92674e0853337b16ee0e2&vid=5b566036df87bda7bbe92674e0853337b16ee0e2&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 99.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR