α-Chlorothioanisole ≥95.0%

Leverantör: TCI
Varning

Synonyms: Chloromethyl phenyl sulphide

C0912-25G C0912-5G
TCIAC0912-25GEA 1850 SEK
TCIAC0912-25G TCIAC0912-5G
α-Chlorothioanisole ≥95.0%
α-Chlorothioanisole
Formel: C₇H₇ClS
Flampunkt: 101°C
Storage Temperature: Kylskåp
MDL Number: MFCD00000921
CAS nummer: 7205-91-6
EINECS: 230-579-7
UN: 3532
ADR: 4.1 III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=de7ff734a3917194c515b4b2ce8dd0e3f621c3f4&vid=8bb12ff723525fd3d6a3d7459beb16c42467c5eb&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 95.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR