Di-n-octyl phthalate ≥98%
Leverantör: Thermo Fisher Scientific
41522.AC 41522.AE 41522.AP
41522.ACEA 1475 SEK
41522.AC 41522.AE 41522.AP
Di-n-octyl phthalate ≥98%
Di-n-octyl phthalate
Purity: 98% CAS No.: 117-84-0 Molecular Formula: C24H38O4 Formula Weight: 390.56 Flash Point: 219°(426°F) Physical Form: Liquid Density: 0.98 Appearance: Colourless to plae yellow Melting Point: -25° Boiling Point: 380° Merck Reference: 14,2864 MDL No.: MFCD00015292
Plasticizer
Formel: C₂₄H₃₈O₄
MW: 390.56 g/mol
Kokpunkt: 380 °C
Smältpunkt: –25 °C
Densitet: 0.98
Flampunkt: 219 °C (426 °F)
Storage Temperature: Omgivande
MDL Number: MFCD00015292
CAS nummer: 117-84-0
EINECS: 204-214-7