Methyl-3-methoxypropionate ≥99.0%

Leverantör: TCI EUROPE
Varning

Synonyms: Methyl 3-methoxypropionate

M0962-25ML M0962-500ML
TCIAM0962-25MLEA 215 SEK
TCIAM0962-25ML TCIAM0962-500ML
Methyl-3-methoxypropionate ≥99.0%
Methyl-3-methoxypropionate
Formel: C₅H₁₀O₃
MW: 118,1311 g/mol
Kokpunkt: 144°C
Flampunkt: 47°C
MDL Number: MFCD00059085
CAS nummer: 3852-09-3
EINECS: 223-358-1
UN: 3272
ADR: 3,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4921ee19f02ed988eba9acce3c8012709fe097b6&vid=4921ee19f02ed988eba9acce3c8012709fe097b6&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 99.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR