Ethyl-3-methoxypropionate ≥99.0%

Leverantör: TCI EUROPE
Varning

Synonyms: 3-Methoxypropionic acid ethyl ester

M1001-10G M1001-25G
TCIAM1001-10GEA 485 SEK
TCIAM1001-10G TCIAM1001-25G
Ethyl-3-methoxypropionate ≥99.0%
Ethyl-3-methoxypropionate
Formel: C₆H₁₂O₃
Kokpunkt: 158°C
MDL Number: MFCD00143172
CAS nummer: 10606-42-5
UN: 3272
ADR: 3,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=609311ee7e93d4e3dad01fdfd06e104232e9713e&vid=609311ee7e93d4e3dad01fdfd06e104232e9713e&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 99.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR