5-Methyl-2,1,3-benzothiadiazole

Leverantör: Thermo Scientific

RDP00067.10
MAYBRDP00067.10EA 0 SEK
MAYBRDP00067.10
5-Methyl-2,1,3-benzothiadiazole
5-Methyl-2,1,3-benzothiadiazole
Formel: C₇H₆N₂S
MDL Number: MFCD00173945
CAS nummer: 1457-93-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR