Laboratory glass cleaner, deconex® 20 NS-x
Leverantör: Borer Chemie
519100.00-F10W 519100.00-K10W 519100.00-KK5W
DECO519100.00-F10WEA 435 SEK
DECO519100.00-F10W DECO519100.00-K10W DECO519100.00-KK5W
Laboratory glass cleaner, deconex® 20 NS-x
Rengöringsmedel Tvätt- och rengöringsmedel
Liquid, alkaline cleaning concentrate for manual and automated cleaning of laboratory glassware und reusable laboratory material.
  • Rinsable without residues and guarantees therefore consistent and error-free results
  • With a broad cleaning spectrum and excellent results even at low concentrations
  • High cleaning performance due to excellent swelling and dispersing capabilities
  • For a residue-free and non-foaming cleaning of delicate items
  • Suitable for soaking baths, ultrasonic devices and cleaning in washing machines
  • surfactant-free, no release of surfactants into waste water
  • EDTA-, NTA- and chlorine-free
  • Rinsing with an acidic deconex® cleaner is recommended as the completion of the automated cleaning process