Thiophene-2-methylamine ≥98.0% (by GC, titration analysis)
Leverantör: TCI EUROPE
Fara
Synonyms: 2-Thiophenemethylamine, 2-(Aminomethyl)thiophene
A2419-25G A2419-5G
TCIAA2419-25GEA 2875 SEK
TCIAA2419-25G TCIAA2419-5G
Thiophene-2-methylamine ≥98.0% (by GC, titration analysis)
Thiophene-2-methylamine
Formel: C₅H₇NS
MW: 113,19 g/mol
Kokpunkt: 99°C
Flampunkt: 74°C
MDL Number: MFCD00005460
CAS nummer: 27757-85-3
EINECS: 248-639-6
UN: 2735
ADR: 8,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5da646080a84d548f21dc145d13faf6bc886b64a&vid=5da646080a84d548f21dc145d13faf6bc886b64a&prid=web&clid=SAPDAM
Order Now
Specifikation testresultat
Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %