Thiophene-2-methylamine 95%
Leverantör: Thermo Scientific
Fara
Synonyms: 2-Thiophenemethylamine, 2-(Aminomethyl)thiophene
205570250 205571000
ACRO205570250EA 2625 SEK
ACRO205570250 ACRO205571000
Thiophene-2-methylamine 95%
Thiophene-2-methylamine
Formel: C₅H₇NS
MW: 113,19 g/mol
Kokpunkt: 95…99 °C (28 mmHg)
Flampunkt: 73 °C
Storage Temperature: Omgivande
MDL Number: MFCD00005460
CAS nummer: 27757-85-3
EINECS: 248-639-6
UN: 2735
ADR: 8,II
Order Now
Specifikation testresultat
Appearance Clear colorless to yellow Liquid
Infrared spectrum Conforms
GC ≥94 %