2-Mercaptopyridine-N-oxide sodium salt hydrate 98%
Leverantör: Thermo Scientific
Varning
Synonyms: N-Hydroxy-2-pyridinethione, sodium salt hydrate, 2-Pyridinethiol-1-oxide, sodium salt hydrate, Sodium omadine hydrate
255380050 255380250
ACRO255380050EA 670 SEK
ACRO255380050 ACRO255380250
2-Mercaptopyridine-N-oxide sodium salt hydrate 98%
2-Mercaptopyridine-N-oxide sodium salt hydrate
Appearance: Off-white to light yellow Powder or granules and/or chunks
Formel: C₅H₄NNaOS
MW: 149,14 g/mol (anhy)
Smältpunkt: >260 °C
Storage Temperature: Omgivande
MDL Number: MFCD00151244
CAS nummer: 3811-73-2
EINECS: 223-296-5
UN: 3077
ADR: 9,III
Order Now
Specifikation testresultat
Appearance Off-white to light yellow Powder or granules and/or chunks
Infrared spectrum Conforms
Loss on drying ≤12 %
Titration with HClO4 ≥97.5 % (On dried substance)