Cyclopropylboronic acid pinacol ester ≥95.0%

Leverantör: TCI
Varning

C2276-1G C2276-5G
TCIAC2276-1GEA 1025 SEK
TCIAC2276-1G TCIAC2276-5G
Cyclopropylboronic acid pinacol ester ≥95.0%
Cyclopropylboronic acid pinacol ester
Formel: C₉H₁₇BO₂
Flampunkt: 40°C
MDL Number: MFCD05663847
CAS nummer: 126689-01-8
UN: 3272
ADR: 3,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0d0f7e55382e10aa96ccd980097a7e781808f1fe&vid=0d0f7e55382e10aa96ccd980097a7e781808f1fe&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 95.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR