5-Cyclohexyl-o-anisidine ≥98.0%
Leverantör: TCI
C3656-10MG
C3656-50MG
TCIAC3656-10MGEA
1100
SEK
TCIAC3656-10MG
TCIAC3656-50MG
5-Cyclohexyl-o-anisidine ≥98.0%
5-Cyclohexyl-o-anisidine
Formel:
C₁₃H₁₉NO Storage Temperature: Frys |
MDL Number:
MFCD00270102 CAS nummer: 206559-52-6 |
Specifikation testresultat
Appearance | White to Yellow to Orange powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Melting point | 173.0 to 177.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...