Benzo[b]thiophene-2-carboxaldehyde

Leverantör: Thermo Scientific

Synonyms: Benzo[b]thiophene-2-carbaldehyde

CC12204DA CC12204EA
MAYBCC12204DAEA 0 SEK
MAYBCC12204DA MAYBCC12204EA
Benzo[b]thiophene-2-carboxaldehyde
Benzo[b]thiophene-2-carboxaldehyde
Formel: C₉H₆OS
MW: 162,21 g/mol
MDL Number: MFCD01075041
CAS nummer: 3541-37-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR