4-(Trifluoromethyl)benzoic acid hydrazide ≥98%
Leverantör: Thermo Fisher Scientific
Varning
Synonyms: 4-(Trifluoromethyl)benzoylhydrazine, 4-(Trifluoromethyl)benzhydrazide, 4-(Trifluoromethyl)benzoic Hydrazide, 4-(Trifluoromethyl)benzohydrazide
L05724.03 L05724.06
L05724.03EA 575 SEK
L05724.03 L05724.06
4-(Trifluoromethyl)benzoic acid hydrazide ≥98%
4-(Trifluoromethyl)benzoic acid hydrazide
Purity: 98% CAS No.: 339-59-3 Molecular Formula: C8H7F3N2O Formula Weight: 204.15 Melting Point: 117-121° Beilstein Reference: 1968848
Formel: C₈H₇F₃N₂O
MW: 204.15 g/mol
Smältpunkt: 117…121 °C
Storage Temperature: Omgivande
MDL Number: MFCD00051703
CAS nummer: 339-59-3