3-(Hydroxymethyl)phenylboronic acid (contains varying amounts of Anhydride)
Leverantör: TCI EUROPE
|
Varning
|
Synonyms:
3-(Hydroxymethyl)benzeneboronic acid, (3-Hydroxymethyl)phenylboronic acid
H1244-1G
H1244-5G
TCIAH1244-1GEA
215
SEK
TCIAH1244-1G
TCIAH1244-5G
3-(Hydroxymethyl)phenylboronic acid (contains varying amounts of Anhydride)
3-(Hydroxymethyl)phenylboronic acid
Formel:
C₇H₉BO₃ MW: 151,96 g/mol |
MDL Number:
MFCD01317846 CAS nummer: 87199-15-3 |
Specifikation testresultat
Appearance | White to Almost white powder to crystal |
Purity (Neutralization titration) | 97.0 to 113.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...