2-(Hydroxymethyl)phenylboronic acid (contains varying amounts of Anhydride)
Leverantör: TCI EUROPE
|
Varning
|
Synonyms:
(2-(Hydroxymethyl)phenyl)boronic acid, 2-Hydroxymethylphenylboronic acid, 2-(Hydroxymethyl)benzeneboronic acid
H1345-1G
H1345-5G
TCIAH1345-1GEA
495
SEK
TCIAH1345-1G
TCIAH1345-5G
2-(Hydroxymethyl)phenylboronic acid (contains varying amounts of Anhydride)
2-(Hydroxymethyl)phenylboronic acid
Formel:
C₇H₉BO₃ MW: 151,96 g/mol |
MDL Number:
MFCD02258966 CAS nummer: 87199-14-2 |
Specifikation testresultat
Appearance | White to Almost white powder to crystal |
Purity (Neutralization titration) | 97.0 to 113.5 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...