4-Nitrophenylsulphur pentafluoride ≥96.0%
Leverantör: TCI EUROPE
|
Fara
|
N0743-1G
N0743-5G
TCIAN0743-1GEA
590
SEK
TCIAN0743-1G
TCIAN0743-5G
4-Nitrophenylsulphur pentafluoride ≥96.0%
4-Nitrophenylsulphur pentafluoride
Formel:
C₆H₄F₅NO₂S MW: 249,16 g/mol Kokpunkt: 77°C |
MDL Number:
MFCD00221619 CAS nummer: 2613-27-6 EINECS: 447-610-7 UN: 2811 ADR: 6.1,III |
Specifikation testresultat
Appearance | Light orange to Yellow to Green powder to crystal |
Purity (GC) | min. 96.0 % |
Melting point | 37.0 to 41.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...