2,4-Diaminoanisole dihydrochloride ≥99.0%

Leverantör: TCI EUROPE
Varning

Synonyms: 4-Methoxybenzene-1,3-diamine dihydrochloride

D0074-25G D0074-5G
TCIAD0074-25GEA 4725 SEK
TCIAD0074-25G TCIAD0074-5G
2,4-Diaminoanisole dihydrochloride ≥99.0%
2,4-Diaminoanisole dihydrochloride
Formel: C₇H₁₂Cl₂N₂O
MDL Number: MFCD00054337
CAS nummer: 614-94-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5dbae50b4d1f9c6c6dee40434c924168d5ba9442&vid=5dbae50b4d1f9c6c6dee40434c924168d5ba9442&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance White to Light yellow to Dark green powder to crystal
Purity (Neutralization titration) min. 99.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR