2-Amino-5-methoxybenzenesulphonic acid ≥97.0%

Leverantör: TCI

A1285-25G A1285-500G
TCIAA1285-25GEA 405 SEK
TCIAA1285-25G TCIAA1285-500G
2-Amino-5-methoxybenzenesulphonic acid ≥97.0%
2-Amino-5-methoxybenzenesulphonic acid
Formel: C₇H₉NO₄S
MDL Number: MFCD00035804
CAS nummer: 13244-33-2
EINECS: 236-224-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ce6c5f203a824cbadfbd687fb1268f4c6b5435b2&vid=ce6c5f203a824cbadfbd687fb1268f4c6b5435b2&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance White to Gray to Brown powder to crystal
Purity (Neutralization titration) min. 97.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR