4-Iodophenylsulphur pentafluoride ≥94.0%
Leverantör: TCI EUROPE
Varning
I0695-1G I0695-5G
TCIAI0695-1GEA 2400 SEK
TCIAI0695-1G TCIAI0695-5G
4-Iodophenylsulphur pentafluoride ≥94.0%
4-Iodophenylsulphur pentafluoride
Formel: C₆H₄F₅IS
MW: 330,600 g/mol
Kokpunkt: 52°C
Storage Temperature: Kylskåp
MDL Number: MFCD03788514
CAS nummer: 286947-68-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d40b96c6741213a0946591200aa4081d5574d1e8&vid=d40b96c6741213a0946591200aa4081d5574d1e8&prid=web&clid=SAPDAM
Order Now
Specifikation testresultat
Appearance Light orange to Yellow to Green powder to crystal
Purity (GC) min. 95.0 %
Melting point 34.0 to 38.0 deg-C