Methyl-5-iodsalicylate ≥98.0%

Leverantör: TCI EUROPE
Varning

Synonyms: Methyl 2-hydroxy-5-iodobenzoate

I0539-25G I0539-5G
TCIAI0539-25GEA 1700 SEK
TCIAI0539-25G TCIAI0539-5G
Methyl-5-iodsalicylate ≥98.0%
Methyl-5-iodsalicylate
Formel: C₈H₇IO₃
MW: 278,05 g/mol
MDL Number: MFCD00016462
CAS nummer: 4068-75-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f3329df3bcabd4494fce9378ca92c32bffdf4de2&vid=f3329df3bcabd4494fce9378ca92c32bffdf4de2&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 75.0 to 78.0 deg-C
Solubility in Methanol very faint turbidity

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR