Methyl-4-iodsalicylate ≥98.0%
Leverantör: TCI EUROPE
|
Varning
|
Synonyms:
Methyl 2-hydroxy-4-iodobenzoate
M1908-25G
M1908-5G
TCIAM1908-25GEA
3375
SEK
TCIAM1908-25G
TCIAM1908-5G
Methyl-4-iodsalicylate ≥98.0%
Methyl-4-iodsalicylate
Formel:
C₈H₇IO₃ MW: 278,05 g/mol |
MDL Number:
MFCD06797864 CAS nummer: 18179-39-0 |
Specifikation testresultat
Appearance | White to Gray to Brown powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 68.0 to 72.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...