Methyl-4-iodsalicylate ≥98%
Leverantör: Thermo Scientific
Varning
Synonyms: Methyl 2-hydroxy-4-iodobenzoate
B25438.14 B25438.06
B25438.14EA 3850 SEK
B25438.14 B25438.06
Methyl-4-iodsalicylate ≥98%
Methyl-4-iodsalicylate
Purity: 98% CAS No.: 18179-39-0 Molecular Formula: C8H7IO3 Formula Weight: 278.05 Melting Point: 68-72° MDL No.: MFCD06797864
Formel: C₈H₇IO₃
MW: 278,05 g/mol
Smältpunkt: 68…72 °C
Storage Temperature: Omgivande
MDL Number: MFCD06797864
CAS nummer: 18179-39-0