Methyl thiosalicylate 97%
Leverantör: Thermo Scientific
Varning
Synonyms: Methyl 2-mercaptobenzoate
369600050 369600250
ACRO369600050EA 1500 SEK
ACRO369600050 ACRO369600250
Methyl thiosalicylate 97%
Methyl thiosalicylate
Formel: C₈H₈O₂S
MW: 168,21 g/mol
Kokpunkt: 98…100 °C (2 mmHg)
Flampunkt: >110 °C
Storage Temperature: Omgivande
MDL Number: MFCD00060678
CAS nummer: 4892-02-8
UN: 2810
ADR: 6.1,III
Order Now
Specifikation testresultat
Appearance Clear colorless to yellow to light green Liquid
Infrared spectrum Conforms
Refractive index 1.5910 to 1.5950 (20°C, 589 nm)
GC ≥96.0 %