Methyl-5-methoxysalicylate ≥98.0%
Leverantör: TCI EUROPE
Varning
Synonyms: Methyl 2-hydroxy-5-methoxybenzoate
M2127-25G M2127-5G
TCIAM2127-25GEA 1825 SEK
TCIAM2127-25G TCIAM2127-5G
Methyl-5-methoxysalicylate ≥98.0%
Methyl-5-methoxysalicylate
Formel: C₉H₁₀O₄
MW: 182,1733 g/mol
Kokpunkt: 240°C
MDL Number: MFCD00017185
CAS nummer: 2905-82-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=69ba0da466d1311333f97619aa2c2cbbad630c7b&vid=69ba0da466d1311333f97619aa2c2cbbad630c7b&prid=web&clid=SAPDAM
Order Now
Specifikation testresultat
Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 98.0 %