4-(Diethylamino)salicylaldehyde ≥98.0% (by GC, titration analysis)
Leverantör: TCI EUROPE
|
Varning
|
Synonyms:
4-(Diethylamino)-2-hydroxybenzaldehyde
D1752-25G
D1752-250G
TCIAD1752-25GEA
535
SEK
TCIAD1752-25G
TCIAD1752-250G
4-(Diethylamino)salicylaldehyde ≥98.0% (by GC, titration analysis)
4-(Diethylamino)salicylaldehyde
Formel:
C₁₁H₁₅NO₂ MW: 193,25 g/mol |
MDL Number:
MFCD00003326 CAS nummer: 17754-90-4 EINECS: 241-745-3 |
Specifikation testresultat
Appearance | White to Brown powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Nonaqueous Titration) | min. 98.0 % |
Melting point | 59.0 to 63.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...