N,N'-Bis(salicylidene)ethylenediamine 98%
Leverantör: Thermo Scientific
|
Varning
|
Synonyms:
Ethylenebis(salicylimine), 2,2'-((Ethane-1,2-diylbis(azanylylidene))bis(methanylylidene))diphenol, SALEN
A13885.14
A13885.22
A13885.36
A13885.14EA
1075
SEK
A13885.14
A13885.22
A13885.36
N,N'-Bis(salicylidene)ethylenediamine 98%
N,N'-Bis(salicylidene)ethylenediamine
Purity: 98%
CAS No.: 94-93-9
Molecular Formula: C16H16N2O2
Formula Weight: 268.32
Melting Point: 129-131°
Beilstein Reference: 535296
Merck Reference: 14,8322
MDL No.: MFCD00002244
Formel:
C₁₆H₁₆N₂O₂ MW: 268,31 g/mol Smältpunkt: 129…131 °C Storage Temperature: Omgivande |
MDL Number:
MFCD00002244 CAS nummer: 94-93-9 EINECS: 202-376-3 Merck Index: 14,8322 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...