9-Ethylcarbazole ≥97.0%

Leverantör: TCI EUROPE
Varning

Synonyms: 9-Ethyl-9H-carbazole, N-Ethylcarbazole

E0071-25G E0071-500G
TCIAE0071-25GEA 245 SEK
TCIAE0071-25G TCIAE0071-500G
9-Ethylcarbazole ≥97.0%
9-Ethylcarbazole
Formel: C₁₄H₁₃N
MW: 195,27 g/mol
Kokpunkt: 166°C
MDL Number: MFCD00004967
CAS nummer: 86-28-2
EINECS: 201-660-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f035ee4e99d5b7e27916a851ebabc1ed0a19f6dc&vid=f035ee4e99d5b7e27916a851ebabc1ed0a19f6dc&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance White to Gray to Brown powder to crystal
Purity (GC) min. 97.0 %
Melting point 68.0 to 71.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR