2'-(Trifluoromethoxy)acetanilide

Leverantör: Thermo Scientific

TL00689DA TL00689EA
MAYBTL00689DAEA 0 SEK
MAYBTL00689DA MAYBTL00689EA
2'-(Trifluoromethoxy)acetanilide
2'-(Trifluoromethoxy)acetanilide
Formel: C₉H₈F₃NO₂
MDL Number: MFCD00204176
CAS nummer: 3832-55-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR