Sodium glycolate

Leverantör: TCI

G0111-25G G0111-500G
TCIAG0111-25GEA 215 SEK
TCIAG0111-25G TCIAG0111-500G
Sodium glycolate
Sodium glycolate
Formel: C₂H₃NaO₃
MDL Number: MFCD00065358
CAS nummer: 2836-32-0
EINECS: 220-624-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1581651203fed564336e132e1e8029dc39cb229d&vid=1581651203fed564336e132e1e8029dc39cb229d&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance White to Almost white powder to crystal
Purity (Nonaqueous Titration) min. 98.0 %
Solubility in Water almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR