Trimethylsilyl acetate (acetoxytrimethylsilane) ≥97%
Leverantör: Thermo Fisher Scientific
Fara
Synonyms: Trimethylsilyl acetate
B22037.14 B22037.22
B22037.14EA 805 SEK
B22037.14 B22037.22
Trimethylsilyl acetate (acetoxytrimethylsilane) ≥97%
Trimethylsilyl acetate (acetoxytrimethylsilane)
Purity: 97% CAS No.: 2754-27-0 Molecular Formula: C5H12O2Si Formula Weight: 132.24 Flash Point: 4°(39°F) Refractive Index: 1.3880 Melting Point: -32° Boiling Point: 107° Beilstein Reference: 1744463 UN No.: 3272 MDL No.: MFCD00008695
Formel: C₅H₁₂O₂Si
Kokpunkt: 107 °C
Smältpunkt: –32 °C
Densitet: 0,884
Flampunkt: 4 °C (39 °F)
Storage Temperature: Omgivande
MDL Number: MFCD00008695
CAS nummer: 2754-27-0
EINECS: 220-404-2
UN: 3272
ADR: 3,II