4'-Acetoxyacetanilide ≥99.0% (by HPLC, total nitrogen)

Leverantör: TCI
Varning

Synonyms: 4-Acetamidophenyl acetate, Acetaminophen Acetate (Acetaminophen Impurity)

A0067-25G A0067-5G
TCIAA0067-25GEA 2400 SEK
TCIAA0067-25G TCIAA0067-5G
4'-Acetoxyacetanilide ≥99.0% (by HPLC, total nitrogen)
4'-Acetoxyacetanilide
Formel: C₁₀H₁₁NO₃
MW: 193.1992 g/mol
MDL Number: MFCD00059205
CAS nummer: 2623-33-8
EINECS: 220-077-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0c23d45862b6b43dbe000ba939df9202e893a8e8&vid=0c23d45862b6b43dbe000ba939df9202e893a8e8&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance White to Light yellow powder to crystal
Purity (HPLC) min. 99.0 area%
Purity (with Total Nitrogen) min. 99.0 %
Melting point 153.0 to 157.0 deg-C
Solubility in Methanol almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR