3-Bromo-1-propylboronic acid pinacol ester ≥98%
Leverantör: Thermo Fisher Scientific
Varning
Synonyms: 3-Bromopropylboronic acid pinacol ester, 2-(3-Bromopropyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
H30646.03 H30646.06 H30646.14
H30646.03EA 1225 SEK
H30646.03 H30646.06 H30646.14
3-Bromo-1-propylboronic acid pinacol ester ≥98%
3-Bromo-1-propylboronic acid pinacol ester
Purity: 98% CAS No.: 124215-44-7 Molecular Formula: C9H18BBrO2 Formula Weight: 248.96 Boiling Point: 61-64°/0.35mm MDL No.: MFCD10567053
Formel: C₉H₁₈BBrO₂
Kokpunkt: 61…64 °C (0,35 mmHg)
Densitet: 1,177
Storage Temperature: Omgivande
MDL Number: MFCD10567053
CAS nummer: 124215-44-7