D-(-)-Penicillamine ≥98.0%

Leverantör: TCI
Varning

P0147-25G P0147-5G
TCIAP0147-25GEA 5225 SEK
TCIAP0147-25G TCIAP0147-5G
D-(-)-Penicillamine ≥98.0%
D-(-)-Penicillamine
Formel: C₅H₁₁NO₂S
MDL Number: MFCD00064302
CAS nummer: 52-67-5
EINECS: 200-148-8
Merck Index: 14,07088
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=87be19f70cf6b6bf659207423afeb02a491a9f94&vid=87be19f70cf6b6bf659207423afeb02a491a9f94&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance White to Almost white powder to crystal
Purity (Neutralization titration) min. 98.0 %
Specific rotation [a]20/D -61.0 to -65.0 deg (C=1, 1mol/L NaOH sol.)
Solubility in Water almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR